Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATXN7L3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098833.1 | NP_001092303.1 | 12 | 2710 | ||
NM_020218.1 | NP_064603.1 | 12 | 2731 | |||
XM_011525038.2 | XP_011523340.1 | 12 | 3382 | |||
XM_011525044.2 | XP_011523346.1 | 12 | 3325 | |||
XM_011525046.2 | XP_011523348.1 | 12 | 3304 | |||
XM_017024880.1 | XP_016880369.1 | 13 | 2960 | |||
XM_017024881.1 | XP_016880370.1 | 13 | 3259 | |||
XM_017024882.1 | XP_016880371.1 | 12 | 3361 | |||
XM_017024883.1 | XP_016880372.1 | 13 | 2963 | |||
XM_017024884.1 | XP_016880373.1 | 12 | 2985 | |||
XM_017024885.1 | XP_016880374.1 | 13 | 2906 | |||
XM_017024886.1 | XP_016880375.1 | 13 | 2885 | |||
GenBank mRNA | AK056002.1 | - | 9 | 2254 | ||
AK074344.1 | - | 1 | 1319 | |||
AL390158.1 | - | 7 | 2138 | |||
BC024695.2 | - | 5 | 2068 | |||
BC037418.1 | - | 12 | 2638 | |||
BC100295.1 | - | 1 | 489 |