Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other FAM122B SIRNA Assays ›
Gene Symbol:
FAM122B
Gene Name:
family with sequence similarity 122B
Gene Aliases:
SPACIA2
Chromosome Location:
Chr.X: 134769566 - 134797232 on Build GRCh38
UniGene ID:
Hs.404706
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s46149
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: FAM122B
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001166599.2
NP_001160071.1
8
1509
NM_001166600.2
NP_001160072.1
10
878
NM_001170756.1
NP_001164227.1
9
766
NM_001170757.1
NP_001164228.1
9
597
NM_145284.5
NP_660327.2
8
712
XM_006724730.2
XP_006724793.1
9
1625
XM_006724732.2
XP_006724795.1
9
1625
XM_006724733.3
XP_006724796.1
8
1562
XM_011531282.1
XP_011529584.1
9
1637
XM_011531283.1
XP_011529585.1
9
1634
XM_011531284.1
XP_011529586.1
9
1625
XM_011531285.1
XP_011529587.1
9
1622
XM_011531286.1
XP_011529588.1
8
1580
XM_011531287.1
XP_011529589.1
9
1637
XM_011531288.1
XP_011529590.1
8
1562
XM_011531289.1
XP_011529591.1
9
1634
XM_011531290.1
XP_011529592.1
8
1565
XM_011531291.1
XP_011529593.1
9
1637
XM_011531292.1
XP_011529594.1
9
1634
XM_011531293.1
XP_011529595.1
9
1622
XM_011531294.1
XP_011529596.1
8
1565
XM_017029306.1
XP_016884795.1
8
1580
XM_017029307.1
XP_016884796.1
8
1577
GenBank mRNA
AB541014.1
-
9
619
AB541015.1
-
9
616
AK124650.1
-
10
851
AK124940.1
-
8
498
AK125991.1
-
9
669
AK292517.1
-
8
631
AK297720.1
-
9
597
AL832218.1
-
8
642
BC019221.2
-
8
1488
3841054
BC032419.2
-
4
285
BC110846.1
-
8
1545
6046376
BX538218.1
-
9
989
Back To Top
More Information
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.31.0-2025.06.23-1.0