Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other FAM122C SIRNA Assays ›
Gene Symbol:
FAM122C
Gene Name:
family with sequence similarity 122C
Gene Aliases:
-
Chromosome Location:
Chr.X: 134796413 - 134855137 on Build GRCh38
UniGene ID:
Hs.269127
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s46151
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: FAM122C
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001170779.1
NP_001164250.1
2
566
NM_001170780.1
NP_001164251.1
5
473
NM_001170782.1
NP_001164253.1
3
386
NM_001170783.1
NP_001164254.1
2
566
NM_001170784.1
NP_001164255.1
3
386
NM_138819.3
NP_620174.1
2
566
XM_005262382.1
XP_005262439.1
3
344
XM_005262383.2
XP_005262440.1
3
344
XM_005262384.4
XP_005262441.1
3
344
XM_005262386.3
XP_005262443.1
3
344
XM_005262387.3
XP_005262444.1
3
344
XM_006724734.3
XP_006724797.1
3
432
XM_006724735.1
XP_006724798.1
3
344
XM_006724736.2
XP_006724799.1
3
344
XM_011531295.1
XP_011529597.1
3
344
XM_011531296.2
XP_011529598.1
3
344
XM_011531297.2
XP_011529599.1
3
344
XM_011531298.2
XP_011529600.1
3
344
XM_011531305.2
XP_011529607.1
3
344
XM_011531306.2
XP_011529608.1
3
344
XM_017029308.1
XP_016884797.1
3
344
XM_017029311.1
XP_016884800.1
3
422
GenBank mRNA
AK295485.1
-
3
324
AK307219.1
-
5
473
AK310079.1
-
2
522
BC017868.1
-
2
565
4699951
BC063491.1
-
2
506
5229041
BC065225.1
-
3
291
5724414
BG532414.1
-
2
563
DB458889.1
-
3
387
Back To Top
More Information
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.31.0-2025.06.23-1.0