Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UGT3A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168316.1 | NP_001161788.1 | 2 | 274 | ||
NM_174914.3 | NP_777574.2 | 3 | 376 | |||
NR_031764.1 | 3 | 376 | ||||
XM_011513988.2 | XP_011512290.1 | 4 | 418 | |||
XM_017009150.1 | XP_016864639.1 | 3 | 427 | |||
GenBank mRNA | AK075383.1 | - | 3 | 337 | ||
AK300751.1 | - | 2 | 235 | |||
AK303154.1 | - | 3 | 337 | |||
AY358416.1 | - | 3 | 311 | |||
BC103924.1 | - | 3 | 325 | 40007344 | ||
BC103925.1 | - | 3 | 325 | 40007348 | ||
DB110369.1 | - | 3 | 377 |