Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other EPB41 SIRNA Assays ›
          Gene Symbol:
          EPB41
          Gene Name:
          erythrocyte membrane protein band 4.1
          Gene Aliases:
          4.1R, EL1, HE
          Chromosome Location:
          Chr.1: 28887091 - 29120046 on Build GRCh38
          UniGene ID:
          Hs.175437
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s4703

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: EPB41
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001166005.1 NP_001159477.1 10 1509
          NM_001166006.1 NP_001159478.1 10 1582
          NM_001166007.1 NP_001159479.1 10 1492
          NM_004437.3 NP_004428.1 11 1572
          NM_203342.2 NP_976217.1 11 1589
          NM_203343.2 NP_976218.1 9 1404
          XM_005245753.1 XP_005245810.1 10 1584
          XM_005245757.1 XP_005245814.1 10 1584
          XM_005245760.1 XP_005245817.1 10 1584
          XM_005245761.1 XP_005245818.1 10 1584
          XM_005245763.1 XP_005245820.1 10 1584
          XM_005245764.1 XP_005245821.1 10 1584
          XM_005245765.1 XP_005245822.1 10 1584
          XM_005245768.1 XP_005245825.1 10 1584
          XM_005245769.1 XP_005245826.1 10 1584
          XM_005245770.1 XP_005245827.1 10 1584
          XM_005245772.4 XP_005245829.1 10 1584
          XM_005245773.4 XP_005245830.1 10 1584
          XM_005245774.1 XP_005245831.1 10 1584
          XM_006710434.1 XP_006710497.1 10 1584
          XM_006710439.1 XP_006710502.1 10 1584
          XM_011540956.1 XP_011539258.1 10 1584
          XM_011540957.1 XP_011539259.1 10 1584
          XM_011540958.1 XP_011539260.1 10 1584
          XM_011540959.1 XP_011539261.1 10 1584
          XM_011540960.1 XP_011539262.1 10 1584
          XM_011540961.1 XP_011539263.1 10 1584
          XM_011540962.1 XP_011539264.1 10 1584
          XM_011540963.2 XP_011539265.1 10 1584
          XM_011540964.1 XP_011539266.1 10 1480
          XM_011540965.2 XP_011539267.1 10 1584
          XM_017000581.1 XP_016856070.1 9 1479
          XM_017000582.1 XP_016856071.1 9 1479
          XM_017000583.1 XP_016856072.1 10 1584
          XM_017000584.1 XP_016856073.1 10 1875
          XM_017000585.1 XP_016856074.1 10 1872
          XM_017000586.1 XP_016856075.1 10 1584
          XM_017000587.1 XP_016856076.1 10 1584
          XM_017000588.1 XP_016856077.1 10 1584
          XM_017000589.1 XP_016856078.1 9 1479
          XM_017000590.1 XP_016856079.1 9 1479
          XM_017000591.1 XP_016856080.1 10 1584
          XM_017000592.1 XP_016856081.1 10 1584
          XM_017000593.1 XP_016856082.1 10 1480
          XM_017000594.1 XP_016856083.1 10 1480
          XM_017000595.1 XP_016856084.1 10 1480
          XM_017000596.1 XP_016856085.1 10 1480
          XM_017000597.1 XP_016856086.1 10 1480
          XM_017000598.1 XP_016856087.1 10 1480
          XM_017000599.1 XP_016856088.1 10 1480
          XM_017000600.1 XP_016856089.1 10 1480
          XM_017000601.1 XP_016856090.1 10 1480
          XM_017000602.1 XP_016856091.1 10 1480
          XM_017000603.1 XP_016856092.1 10 1480
          XM_017000604.1 XP_016856093.1 10 1480
          GenBank mRNA AB209649.1 - 10 1492
          AF156225.1 - 9 1382
          AK310655.1 - 9 1420
          BC039079.1 - 10 1579 5167140
          BC096104.1 - 7 798
          BC096105.1 - 9 1116 40001729
          BC113841.1 - 9 1116
          BC114450.1 - 9 1116
          J03796.1 - 9 1323
          M14993.1 - 11 1553
          M61733.1 - 11 1570

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          blood circulation
          actin cytoskeleton organization
          cortical actin cytoskeleton organization
          positive regulation of protein binding
          actin binding
          structural constituent of cytoskeleton
          protein binding
          calmodulin binding
          1-phosphatidylinositol binding
          spectrin binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0