Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TMEM218 SIRNA Assays ›
Gene Symbol:
TMEM218
Gene Name:
transmembrane protein 218
Gene Aliases:
-
Chromosome Location:
Chr.11: 125094370 - 125111763 on Build GRCh38
UniGene ID:
Hs.731899
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s47733
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: TMEM218
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001080546.2
NP_001074015.1
4
613
NM_001258238.1
NP_001245167.1
4
491
NM_001258239.1
NP_001245168.1
4
588
NM_001258240.1
NP_001245169.1
6
651
NM_001258241.1
NP_001245170.1
5
592
NM_001258242.1
NP_001245171.1
4
579
NM_001258243.1
NP_001245172.1
5
682
NM_001258244.1
NP_001245173.1
4
482
NM_001258245.1
NP_001245174.1
6
644
NM_001258246.1
NP_001245175.1
4
564
NM_001258247.1
NP_001245176.1
4
467
NR_047587.1
2
227
NR_047589.1
3
296
NR_047591.1
4
355
XM_006718779.3
XP_006718842.1
6
792
XM_006718780.3
XP_006718843.1
5
689
XM_006718781.3
XP_006718844.1
6
746
XM_006718783.3
XP_006718846.1
4
555
XM_017017322.1
XP_016872811.1
5
579
XM_017017323.1
XP_016872812.1
4
505
XM_017017324.1
XP_016872813.1
4
507
GenBank mRNA
AK091324.1
-
1
3790
AL833215.1
-
1
194
BC132716.1
-
4
450
40146738
BC132718.1
-
4
450
40146740
BC144270.1
-
5
553
9052790
BC144277.1
-
4
484
9052797
BE513148.1
-
4
423
BE974033.1
-
2
72
BI547955.1
-
4
241
BI829527.1
-
4
426
BI856825.1
-
4
357
BM042315.1
-
5
443
BM928068.1
-
523
BQ223298.1
-
6
540
BQ225039.1
-
3
213
BQ420663.1
-
4
458
BQ653243.1
-
4
458
BQ681518.1
-
5
441
BU855096.1
-
4
379
DA899308.1
-
4
451
Back To Top
More Information
Gene Ontology Categories:
Process(es)
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7699c4d8ff-7jgns:80/100.66.130.62:80.
git-commit: c11338b6b8d815146c5ef25758b63739976b60cb
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.31.0-Offline