Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ZNF438 SIRNA Assays ›
Gene Symbol:
ZNF438
Gene Name:
zinc finger protein 438
Gene Aliases:
bA330O11.1
Chromosome Location:
Chr.10: 30844634 - 31032406 on Build GRCh38
UniGene ID:
Hs.660642
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s47913
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ZNF438
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001143766.1
NP_001137238.1
7
1460
NM_001143767.1
NP_001137239.1
5
1273
NM_001143768.1
NP_001137240.1
6
1331
NM_001143769.1
NP_001137241.1
7
1585
NM_001143770.1
NP_001137242.1
8
1511
NM_001143771.1
NP_001137243.1
7
1482
NM_182755.2
NP_877432.1
7
1412
NR_026560.1
6
495
XM_006717393.3
XP_006717456.1
5
1227
XM_006717396.3
XP_006717459.1
4
1161
XM_006717398.3
XP_006717461.1
8
1452
XM_006717399.3
XP_006717462.1
7
1385
XM_011519371.2
XP_011517673.1
6
3435
XM_011519372.2
XP_011517674.1
6
1462
XM_011519373.2
XP_011517675.1
5
3377
XM_011519374.2
XP_011517676.1
3
1204
XM_011519376.2
XP_011517678.1
8
1537
XM_011519377.2
XP_011517679.1
8
1533
XM_017015863.1
XP_016871352.1
3
1027
XM_017015864.1
XP_016871353.1
8
3671
XM_017015865.1
XP_016871354.1
7
3612
XM_017015866.1
XP_016871355.1
7
1502
XM_017015867.1
XP_016871356.1
6
3417
XM_017015868.1
XP_016871357.1
5
1723
XM_017015869.1
XP_016871358.1
5
3358
XM_017015870.1
XP_016871359.1
6
2257
XM_017015871.1
XP_016871360.1
5
1717
XM_017015872.1
XP_016871361.1
6
3415
XM_017015873.1
XP_016871362.1
7
3592
XM_017015874.1
XP_016871363.1
5
1278
XM_017015875.1
XP_016871364.1
7
481
XM_017015876.1
XP_016871365.1
9
1230
XM_017015877.1
XP_016871366.1
8
660
XM_017015878.1
XP_016871367.1
7
587
XM_017015879.1
XP_016871368.1
7
667
XM_017015880.1
XP_016871369.1
6
470
XM_017015881.1
XP_016871370.1
5
408
XM_017015882.1
XP_016871371.1
6
515
GenBank mRNA
AF428258.1
-
6
1307
AK057323.1
-
6
483
AK131357.1
-
7
1585
AK292730.1
-
7
1378
AK294253.1
-
7
471
AL833056.1
-
2
1021
BC101622.1
-
3
1037
8069128
BC104757.1
-
3
1037
8143760
BC143462.1
-
4
1136
BC143464.1
-
4
1216
DQ356011.1
-
7
1406
DQ356012.1
-
7
1397
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
regulation of transcription, DNA-templated
DNA binding
metal ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.31.0-2025.06.23-1.0