Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other C1ORF101 SIRNA Assays ›
Gene Symbol:
C1orf101
Gene Name:
chromosome 1 open reading frame 101
Gene Aliases:
-
Chromosome Location:
Chr.1: 244452014 - 244640362 on Build GRCh38
UniGene ID:
Hs.459534
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s48840
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: C1orf101
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001130957.1
NP_001124429.1
10
1236
NM_001242340.1
NP_001229269.1
7
1084
NM_173807.4
NP_776168.1
10
1236
XM_011544138.2
XP_011542440.1
10
1287
XM_011544139.2
XP_011542441.1
9
1182
XM_011544142.2
XP_011542444.1
11
1510
XM_011544144.2
XP_011542446.1
9
1213
XM_011544145.2
XP_011542447.1
8
1107
XM_011544149.2
XP_011542451.1
8
1069
XM_011544151.2
XP_011542453.1
8
1071
XM_017000943.1
XP_016856432.1
10
1288
XM_017000944.1
XP_016856433.1
6
1456
XM_017000945.1
XP_016856434.1
7
1079
XM_017000946.1
XP_016856435.1
9
1387
XM_017000947.1
XP_016856436.1
10
1288
XM_017000948.1
XP_016856437.1
11
1232
XM_017000949.1
XP_016856438.1
9
1437
XM_017000950.1
XP_016856439.1
8
1083
XM_017000951.1
XP_016856440.1
10
1288
XM_017000952.1
XP_016856441.1
10
1288
XM_017000953.1
XP_016856442.1
10
1289
GenBank mRNA
AK027205.1
-
7
723
AK302609.1
-
10
1189
AK303059.1
-
7
1044
BC032859.1
-
10
1221
5269307
BX648685.1
-
6
937
Back To Top
More Information
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.33.0-2025.07.30-1.0