Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SCGB2B2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025591.3 | NP_001020762.1 | 2 | 213 | ||
GenBank mRNA | AK093495.1 | - | 3 | 2278 | ||
BC093909.1 | - | 2 | 213 | 7939754 | ||
BC093935.1 | - | 2 | 213 | 7939780 | ||
BX105421.1 | - | 1 | 51 |