Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF789 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013258.1 | NP_001013276.1 | 3 | 358 | ||
NM_213603.2 | NP_998768.2 | 3 | 358 | |||
XM_011516067.2 | XP_011514369.1 | 4 | 514 | |||
XM_011516070.2 | XP_011514372.1 | 2 | 352 | |||
XM_011516073.2 | XP_011514375.1 | 1 | 70 | |||
XM_017012018.1 | XP_016867507.1 | 3 | 310 | |||
XM_017012019.1 | XP_016867508.1 | 3 | 360 | |||
XM_017012020.1 | XP_016867509.1 | 4 | 513 | |||
XM_017012021.1 | XP_016867510.1 | 4 | 513 | |||
XM_017012023.1 | XP_016867512.1 | 3 | 430 | |||
XM_017012024.1 | XP_016867513.1 | 3 | 427 | |||
XM_017012025.1 | XP_016867514.1 | 3 | 360 | |||
XM_017012026.1 | XP_016867515.1 | 3 | 428 | |||
XM_017012027.1 | XP_016867516.1 | 4 | 511 | |||
GenBank mRNA | AK093141.1 | - | 3 | 358 | ||
AK309467.1 | - | 4 | 443 | |||
BC031249.1 | - | 4 | 443 | |||
BC089424.1 | - | 3 | 320 | 3054018 | ||
BI767175.1 | - | 2 | 223 | |||
BM840763.1 | - | 3 | 342 |