Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other NEB SIRNA Assays ›
          Gene Symbol:
          NEB
          Gene Name:
          nebulin
          Gene Aliases:
          NEB177D, NEM2
          Chromosome Location:
          Chr.2: 151485334 - 151734487 on Build GRCh38
          UniGene ID:
          Hs.588655
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s502573

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: NEB
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001164507.1 NP_001157979.1 5 487
          NM_001164508.1 NP_001157980.1 5 487
          NM_001271208.1 NP_001258137.1 5 487
          NM_004543.4 NP_004534.2 5 487
          XM_005246590.2 XP_005246647.1 5 474
          XM_005246591.2 XP_005246648.1 5 474
          XM_005246592.2 XP_005246649.1 5 474
          XM_005246593.2 XP_005246650.1 5 474
          XM_005246594.2 XP_005246651.1 5 474
          XM_005246596.2 XP_005246653.1 5 474
          XM_005246597.2 XP_005246654.1 5 474
          XM_005246598.2 XP_005246655.1 5 474
          XM_005246599.2 XP_005246656.1 5 474
          XM_005246600.2 XP_005246657.1 5 474
          XM_005246601.2 XP_005246658.1 5 474
          XM_005246602.2 XP_005246659.1 5 474
          XM_005246603.2 XP_005246660.1 5 474
          XM_005246604.2 XP_005246661.1 5 474
          XM_005246606.2 XP_005246663.1 5 474
          XM_005246608.2 XP_005246665.1 5 474
          XM_005246610.2 XP_005246667.1 5 474
          XM_005246611.2 XP_005246668.1 5 474
          XM_005246612.2 XP_005246669.1 5 474
          XM_005246613.2 XP_005246670.1 5 474
          XM_005246615.2 XP_005246672.1 5 474
          XM_005246616.1 XP_005246673.1 5 474
          XM_005246617.2 XP_005246674.1 5 474
          XM_006712541.2 XP_006712604.1 5 474
          XM_006712542.2 XP_006712605.1 5 474
          XM_011511225.2 XP_011509527.1 5 474
          XM_011511226.2 XP_011509528.1 5 474
          XM_011511227.2 XP_011509529.1 5 474
          XM_017004177.1 XP_016859666.1 5 474
          XM_017004178.1 XP_016859667.1 5 474
          XM_017004179.1 XP_016859668.1 5 474
          XM_017004180.1 XP_016859669.1 5 474
          XM_017004181.1 XP_016859670.1 5 474
          XM_017004182.1 XP_016859671.1 5 474
          XM_017004183.1 XP_016859672.1 5 474
          XM_017004184.1 XP_016859673.1 5 474
          XM_017004185.1 XP_016859674.1 5 474
          GenBank mRNA AK096903.1 - 5 474
          AK307607.1 - 5 474
          BC029384.1 - 5 474
          BC056251.1 - 5 478
          BC105590.1 - 5 478
          X83957.1 - 724

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          muscle organ development
          somatic muscle development
          muscle filament sliding
          regulation of actin filament length
          actin binding
          protein binding
          structural constituent of muscle

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5bbdb6fccc-rsc2d:80/100.66.130.62:80.
          git-commit: 26de9a04ea7719c3b15362b12d28d672cac851f3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: feature/2.32.0/CFGMAGR/CFGMAGR-4285/CFGMAGR-4312-Unified_Card_Service_Magellan_Search_Fix