Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF181 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001029997.3 | NP_001025168.2 | 4 | 1694 | ||
NM_001145665.1 | NP_001139137.1 | 4 | 1691 | |||
XM_005258849.2 | XP_005258906.1 | 5 | 1462 | |||
XM_005258850.2 | XP_005258907.1 | 5 | 1459 | |||
XM_005258851.3 | XP_005258908.1 | 5 | 1450 | |||
XM_006723183.3 | XP_006723246.1 | 5 | 1716 | |||
XM_006723184.3 | XP_006723247.1 | 5 | 1447 | |||
XM_017026739.1 | XP_016882228.1 | 5 | 2204 | |||
XM_017026740.1 | XP_016882229.1 | 4 | 1324 | |||
GenBank mRNA | AK295492.1 | - | 5 | 1361 | ||
AK297316.1 | - | 1 | 526 | |||
BC043228.1 | - | 3 | 2335 | 5295602 | ||
BC104759.1 | - | 4 | 1320 | 8143762 | ||
BC143442.1 | - | 4 | 1317 | 9051950 |