Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM220 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004313.1 | NP_001004313.1 | 6 | 908 | ||
XM_005256634.4 | XP_005256691.1 | 5 | 843 | |||
XM_011523835.2 | XP_011522137.1 | 5 | 392 | |||
XM_011523836.2 | XP_011522138.1 | 4 | 784 | |||
GenBank mRNA | AK289624.1 | - | 6 | 564 | ||
AK295928.1 | - | 5 | 573 | |||
AY550194.1 | - | 6 | 908 | |||
BC127705.1 | - | 6 | 472 | 40127270 | ||
BC136838.1 | - | 6 | 474 | 9020829 | ||
BC136840.1 | - | 6 | 474 | 9020831 | ||
EF126147.1 | - | 5 | 400 |