Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other ANK2 SIRNA Assays ›
          Gene Symbol:
          ANK2
          Gene Name:
          ankyrin 2, neuronal
          Gene Aliases:
          ANK-2, LQT4, brank-2
          Chromosome Location:
          Chr.4: 112706083 - 113383740 on Build GRCh38
          UniGene ID:
          Hs.620557
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s531111

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: ANK2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001127493.1 NP_001120965.1 32 3594
          NM_001148.4 NP_001139.3 30 3500
          NM_020977.3 NP_066187.2 30 3500
          XM_005262945.2 XP_005263002.1 29 3499
          XM_017008067.1 XP_016863556.1 32 4457
          XM_017008068.1 XP_016863557.1 33 4698
          XM_017008069.1 XP_016863558.1 31 4432
          XM_017008070.1 XP_016863559.1 32 4643
          XM_017008071.1 XP_016863560.1 33 3626
          XM_017008072.1 XP_016863561.1 32 3602
          XM_017008073.1 XP_016863562.1 30 3598
          XM_017008074.1 XP_016863563.1 32 3572
          XM_017008075.1 XP_016863564.1 31 3536
          XM_017008076.1 XP_016863565.1 30 3598
          XM_017008077.1 XP_016863566.1 29 3413
          XM_017008078.1 XP_016863567.1 30 3598
          XM_017008079.1 XP_016863568.1 28 3328
          XM_017008080.1 XP_016863569.1 30 3449
          XM_017008081.1 XP_016863570.1 29 3499
          XM_017008082.1 XP_016863571.1 31 3676
          XM_017008083.1 XP_016863572.1 31 3739
          XM_017008084.1 XP_016863573.1 31 3634
          XM_017008085.1 XP_016863574.1 31 3595
          XM_017008086.1 XP_016863575.1 30 3496
          XM_017008087.1 XP_016863576.1 32 3775
          XM_017008088.1 XP_016863577.1 33 3569
          XM_017008089.1 XP_016863578.1 30 3742
          XM_017008090.1 XP_016863579.1 31 3739
          XM_017008091.1 XP_016863580.1 29 3436
          XM_017008092.1 XP_016863581.1 31 3536
          XM_017008093.1 XP_016863582.1 32 3533
          XM_017008094.1 XP_016863583.1 31 3595
          XM_017008095.1 XP_016863584.1 29 3413
          XM_017008096.1 XP_016863585.1 33 3569
          XM_017008097.1 XP_016863586.1 30 3374
          XM_017008098.1 XP_016863587.1 31 3536
          XM_017008099.1 XP_016863588.1 32 3533
          XM_017008100.1 XP_016863589.1 29 3338
          XM_017008101.1 XP_016863590.1 29 3499
          XM_017008102.1 XP_016863591.1 28 3314
          XM_017008103.1 XP_016863592.1 31 3595
          XM_017008104.1 XP_016863593.1 30 3437
          XM_017008105.1 XP_016863594.1 29 3413
          XM_017008106.1 XP_016863595.1 28 3400
          XM_017008107.1 XP_016863596.1 30 3374
          XM_017008108.1 XP_016863597.1 28 3298
          XM_017008109.1 XP_016863598.1 29 3398
          XM_017008110.1 XP_016863599.1 30 3437
          XM_017008111.1 XP_016863600.1 29 3413
          XM_017008112.1 XP_016863601.1 27 3215
          GenBank mRNA AK294720.1 - 9 1245
          AK299767.1 - 31 3524
          AK299893.1 - 3 342
          AK307601.1 - 30 3451
          BX537758.1 - 32 3568
          BX538132.1 - 4 437
          CN259048.1 - 3 402
          X56958.2 - 30 3415
          Z26634.2 - 30 3400

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of heart rate
          atrial septum development
          cellular calcium ion homeostasis
          ER to Golgi vesicle-mediated transport
          cytoskeleton organization
          positive regulation of gene expression
          regulation of cardiac muscle contraction by regulation of the release of sequestered calcium ion
          regulation of cardiac muscle contraction by calcium ion signaling
          paranodal junction assembly
          regulation of protein stability
          T-tubule organization
          protein localization to organelle
          protein localization to cell surface
          cellular protein localization
          protein localization to M-band
          protein localization to T-tubule
          positive regulation of potassium ion transport
          protein stabilization
          regulation of release of sequestered calcium ion into cytosol
          response to methylmercury
          regulation of calcium ion transport
          positive regulation of calcium ion transport
          regulation of cardiac muscle contraction
          regulation of ventricular cardiac muscle cell membrane repolarization
          sarcoplasmic reticulum calcium ion transport
          protein localization to endoplasmic reticulum
          protein localization to plasma membrane
          protein targeting to plasma membrane
          regulation of cardiac muscle cell contraction
          ventricular cardiac muscle cell action potential
          atrial cardiac muscle cell action potential
          SA node cell action potential
          membrane depolarization during SA node cell action potential
          atrial cardiac muscle cell to AV node cell communication
          SA node cell to atrial cardiac muscle cell communication
          regulation of heart rate by cardiac conduction
          regulation of SA node cell action potential
          regulation of atrial cardiac muscle cell action potential
          positive regulation of potassium ion transmembrane transporter activity
          regulation of calcium ion transmembrane transporter activity
          positive regulation of calcium ion transmembrane transporter activity
          positive regulation of cation channel activity
          structural constituent of cytoskeleton
          protein binding
          cytoskeletal adaptor activity
          enzyme binding
          protein kinase binding
          spectrin binding
          protein binding, bridging
          ion channel binding
          ATPase binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-867f8cf58d-sfwzd:80/100.66.130.62:80.
          git-commit: 7a7e1ef32845d1da800a461bcf18a29ea3e3afc0
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: feature/2.32.0/CFGMAGR/CFGMAGR-4285/CFGMAGR-4312-Unified_Card_Service_Magellan_Search_Fix