Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other CFP SIRNA Assays ›
          Gene Symbol:
          CFP
          Gene Name:
          complement factor properdin
          Gene Aliases:
          BFD, PFC, PFD, PROPERDIN
          Chromosome Location:
          Chr.X: 47624213 - 47630305 on Build GRCh38
          UniGene ID:
          Hs.53155
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s533605

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: CFP
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001145252.1 NP_001138724.1 2 332
          NM_002621.2 NP_002612.1 3 448
          GenBank mRNA AK122955.1 - 2 257
          AK310695.1 - 2 327
          BC015756.1 - 2 266 4850121
          DB146591.1 - 2 333
          KU178249.1 - 2 206
          KU178250.1 - 2 206
          M83652.1 - 2 278
          X57748.1 - 2 271
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
          GenBankmRNA BI911264.1 - 2 313 Chr. X - 47627149 - 47629920 [-] on Build GRCh38
          GenBankmRNA CD615209.1 - 2 237 Chr. X - 47627576 - 47629875 [-] on Build GRCh38
          GenBankmRNA BP302470.1 - 2 223 Chr. X - 47627482 - 47629861 [-] on Build GRCh38
          GenBankmRNA DN998876.1 - 2 280 Chr. X - 47627483 - 47629912 [-] on Build GRCh38
          GenBankmRNA BI835950.1 - 276 Chr. - - [-] on Build
          GenBankmRNA BP294978.1 - 2 289 Chr. X - 47627548 - 47629927 [-] on Build GRCh38
          GenBankmRNA CD615205.1 - 2 209 Chr. X - 47627290 - 47629847 [-] on Build GRCh38
          GenBankmRNA DA432930.1 - 2 290 Chr. X - 47627574 - 47629928 [-] on Build GRCh38
          GenBankmRNA DA581797.1 - 2 257 Chr. X - 47627539 - 47629895 [-] on Build GRCh38
          GenBankmRNA BI769024.1 - 2 242 Chr. X - 47627534 - 47629881 [-] on Build GRCh38
          GenBankmRNA BQ067280.1 - 271 Chr. - - [-] on Build
          GenBankmRNA HY086586.1 - 2 315 Chr. X - 47628170 - 47629952 [-] on Build GRCh38
          GenBankmRNA BI770026.1 - 2 269 Chr. X - 47627144 - 47629908 [-] on Build GRCh38
          GenBankmRNA BP295571.1 - 2 288 Chr. X - 47627547 - 47629926 [-] on Build GRCh38
          GenBankmRNA DB152988.1 - 2 327 Chr. X - 47627600 - 47629965 [-] on Build GRCh38
          GenBankmRNA HY104470.1 - 2 228 Chr. X - 47628102 - 47629866 [-] on Build GRCh38
          GenBankmRNA DA959891.1 - 2 267 Chr. X - 47627551 - 47629895 [-] on Build GRCh38
          GenBankmRNA DB133165.1 - 2 257 Chr. X - 47629313 - 47629895 [-] on Build GRCh38
          GenBankmRNA DA948139.1 - 2 258 Chr. X - 47627540 - 47629895 [-] on Build GRCh38
          GenBankmRNA BM920180.1 - 237 Chr. - - [-] on Build
          GenBankmRNA BM007993.1 - 1 16 Chr. X - 47626492 - 47629559 [-] on Build GRCh38
          GenBankmRNA BG341989.1 - 134 Chr. - - [-] on Build
          GenBankmRNA BP339567.1 - 2 229 Chr. X - 47627494 - 47629865 [-] on Build GRCh38
          GenBankmRNA BI771667.1 - 2 306 Chr. X - 47627282 - 47629932 [-] on Build GRCh38
          GenBankmRNA CD615211.1 - 2 242 Chr. X - 47627609 - 47629881 [-] on Build GRCh38
          GenBankmRNA BI768416.1 - 212 Chr. - - [-] on Build
          GenBankmRNA BI766222.1 - 2 201 Chr. X - 47626833 - 47629839 [-] on Build GRCh38
          GenBankmRNA BP295572.1 - 2 288 Chr. X - 47627553 - 47629926 [-] on Build GRCh38
          GenBankmRNA BG745320.1 - 2 266 Chr. X - 47627519 - 47629904 [-] on Build GRCh38
          GenBankmRNA BM918830.1 - 216 Chr. - - [-] on Build
          GenBankmRNA HY064642.1 - 2 229 Chr. X - 47627600 - 47629866 [-] on Build GRCh38
          GenBankmRNA BQ709007.1 - 210 Chr. - - [-] on Build
          GenBankmRNA BI909576.1 - 228 Chr. - - [-] on Build
          GenBankmRNA CD615202.1 - 2 259 Chr. X - 47627540 - 47629897 [-] on Build GRCh38
          GenBankmRNA CD615204.1 - 2 209 Chr. X - 47627301 - 47629847 [-] on Build GRCh38
          GenBankmRNA CD615198.1 - 2 195 Chr. X - 47627280 - 47629833 [-] on Build GRCh38
          GenBankmRNA BJ994957.1 - 2 273 Chr. X - 47627535 - 47629911 [-] on Build GRCh38
          GenBankmRNA DA432828.1 - 2 268 Chr. X - 47627553 - 47629906 [-] on Build GRCh38
          GenBankmRNA DB151007.1 - 2 314 Chr. X - 47628122 - 47629952 [-] on Build GRCh38
          GenBankmRNA CV028988.1 - 2 206 Chr. X - 47627482 - 47629844 [-] on Build GRCh38
          GenBankmRNA BI771282.1 - 2 318 Chr. X - 47627259 - 47629957 [-] on Build GRCh38
          GenBankmRNA DA939691.1 - 2 279 Chr. X - 47627539 - 47629917 [-] on Build GRCh38
          GenBankmRNA CD615214.1 - 2 254 Chr. X - 47627281 - 47629886 [-] on Build GRCh38
          GenBankmRNA BP294705.1 - 2 286 Chr. X - 47627543 - 47629924 [-] on Build GRCh38
          GenBankmRNA BG759290.1 - 1 134 Chr. X - 47626885 - 47629677 [-] on Build GRCh38
          GenBankmRNA BQ711791.1 - 218 Chr. - - [-] on Build
          GenBankmRNA BM926583.1 - 2 207 Chr. X - 47626796 - 47629835 [-] on Build GRCh38
          GenBankmRNA BI836039.1 - 2 206 Chr. X - 47626908 - 47629844 [-] on Build GRCh38
          GenBankmRNA BI771749.1 - 2 295 Chr. X - 47627215 - 47629934 [-] on Build GRCh38
          GenBankmRNA BM545062.1 - 2 299 Chr. X - 47626821 - 47629914 [-] on Build GRCh38

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s)

          immune response
          complement activation
          complement activation, alternative pathway
          regulation of complement activation
          protein O-linked fucosylation
          defense response to bacterium

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-9d5985c9f-cd4wl:80/100.66.130.62:80.
          git-commit: 2310f86ea667df8f532fad8d69a2d0b8b05e2a1e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0