Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other IFT122 SIRNA Assays ›
Gene Symbol:
IFT122
Gene Name:
intraflagellar transport 122
Gene Aliases:
CED, CED1, SPG, WDR10, WDR10p, WDR140
Chromosome Location:
Chr.3: 129440036 - 129520507 on Build GRCh38
UniGene ID:
Hs.655284
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s534719
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: IFT122
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001280541.1
NP_001267470.1
11
1323
NM_001280545.1
NP_001267474.1
12
1498
NM_001280546.1
NP_001267475.1
9
1091
NM_018262.3
NP_060732.2
10
1170
NM_052985.3
NP_443711.2
12
1500
NM_052989.2
NP_443715.1
11
1347
NM_052990.2
NP_443716.1
8
1014
XM_005247609.1
XP_005247666.1
10
1135
XM_006713689.1
XP_006713752.1
11
1312
XM_006713691.2
XP_006713754.1
11
1312
XM_006713692.2
XP_006713755.1
10
1135
XM_006713695.2
XP_006713758.1
11
1312
XM_011512972.2
XP_011511274.1
11
1312
XM_017006817.1
XP_016862306.1
11
1312
XM_017006818.1
XP_016862307.1
11
1312
XM_017006819.1
XP_016862308.1
11
1312
XM_017006820.1
XP_016862309.1
11
1312
XM_017006821.1
XP_016862310.1
11
1312
XM_017006822.1
XP_016862311.1
9
1103
XM_017006823.1
XP_016862312.1
10
1135
XM_017006824.1
XP_016862313.1
10
1135
XM_017006825.1
XP_016862314.1
10
1135
XM_017006826.1
XP_016862315.1
10
1135
XM_017006827.1
XP_016862316.1
10
1135
XM_017006828.1
XP_016862317.1
8
926
XM_017006829.1
XP_016862318.1
8
926
XM_017006830.1
XP_016862319.1
12
1310
XM_017006831.1
XP_016862320.1
10
1231
XM_017006832.1
XP_016862321.1
11
1210
XM_017006833.1
XP_016862322.1
8
926
XM_017006834.1
XP_016862323.1
8
926
XM_017006835.1
XP_016862324.1
9
1054
XM_017006836.1
XP_016862325.1
11
1133
XM_017006837.1
XP_016862326.1
2
237
XM_017006838.1
XP_016862327.1
7
677
GenBank mRNA
AF244930.1
-
11
1244
AF244931.1
-
12
1397
AF302154.1
-
11
1129
AK001759.1
-
10
983
AK022525.1
-
10
1081
AK023000.1
-
10
812
AK024435.1
-
9
858
AK122965.1
-
5
965
AK124140.1
-
12
1301
AK223095.1
-
10
982
AK293852.1
-
8
828
AK294515.1
-
6
719
AK298526.1
-
7
591
AK299903.1
-
9
804
AK315103.1
-
11
1147
AL133565.1
-
9
924
BC003045.1
-
8
806
2823536
BC004238.2
-
8
806
2823536
BC028353.2
-
11
1087
4836844
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
neural tube closure
signal transduction downstream of smoothened
embryonic body morphogenesis
negative regulation of smoothened signaling pathway involved in ventral spinal cord patterning
embryonic heart tube development
embryonic forelimb morphogenesis
intraciliary anterograde transport
intraciliary retrograde transport
intraciliary transport involved in cilium morphogenesis
cilium assembly
embryonic digit morphogenesis
negative regulation of smoothened signaling pathway
camera-type eye morphogenesis
negative regulation of epithelial cell proliferation
limb development
cilium morphogenesis
ciliary receptor clustering involved in smoothened signaling pathway
smoothened signaling pathway involved in dorsal/ventral neural tube patterning
embryonic heart tube left/right pattern formation
protein localization to cilium
establishment of protein localization to organelle
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7b8d7ffddb-2cdg5:80/100.66.130.62:80.
git-commit: 44eaffc35a31f29f64b20d0c2dd51bfa522c97f5
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.31.0-2025.06.23-1.0