Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other IFT122 SIRNA Assays ›
          Gene Symbol:
          IFT122
          Gene Name:
          intraflagellar transport 122
          Gene Aliases:
          CED, CED1, SPG, WDR10, WDR10p, WDR140
          Chromosome Location:
          Chr.3: 129440036 - 129520507 on Build GRCh38
          UniGene ID:
          Hs.655284
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s534721

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: IFT122
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001280541.1 NP_001267470.1 16 2145
          NM_001280545.1 NP_001267474.1 17 2320
          NM_001280546.1 NP_001267475.1 14 1913
          NM_018262.3 NP_060732.2 15 1992
          NM_052985.3 NP_443711.2 17 2322
          NM_052989.2 NP_443715.1 16 2169
          NM_052990.2 NP_443716.1 13 1836
          XM_005247609.1 XP_005247666.1 15 1957
          XM_006713689.1 XP_006713752.1 16 2134
          XM_006713691.2 XP_006713754.1 16 2134
          XM_006713692.2 XP_006713755.1 15 1957
          XM_006713695.2 XP_006713758.1 16 2134
          XM_011512972.2 XP_011511274.1 16 2134
          XM_017006817.1 XP_016862306.1 16 2134
          XM_017006818.1 XP_016862307.1 16 2134
          XM_017006819.1 XP_016862308.1 16 2134
          XM_017006820.1 XP_016862309.1 16 2134
          XM_017006821.1 XP_016862310.1 16 2134
          XM_017006822.1 XP_016862311.1 14 1925
          XM_017006823.1 XP_016862312.1 15 1957
          XM_017006824.1 XP_016862313.1 15 1957
          XM_017006825.1 XP_016862314.1 15 1957
          XM_017006826.1 XP_016862315.1 15 1957
          XM_017006827.1 XP_016862316.1 15 1957
          XM_017006828.1 XP_016862317.1 13 1748
          XM_017006829.1 XP_016862318.1 13 1748
          XM_017006830.1 XP_016862319.1 17 2132
          XM_017006831.1 XP_016862320.1 15 2053
          XM_017006832.1 XP_016862321.1 16 2032
          XM_017006833.1 XP_016862322.1 13 1748
          XM_017006834.1 XP_016862323.1 13 1748
          XM_017006835.1 XP_016862324.1 14 1876
          XM_017006836.1 XP_016862325.1 16 1955
          XM_017006837.1 XP_016862326.1 7 1078
          XM_017006838.1 XP_016862327.1 12 1518
          GenBank mRNA AF244930.1 - 16 2066
          AF244931.1 - 17 2219
          AF302154.1 - 16 1951
          AK000773.1 - 1 1189
          AK001759.1 - 15 1805
          AK001955.1 - 5 614
          AK022525.1 - 15 1903
          AK023000.1 - 15 1634
          AK024435.1 - 14 1680
          AK094922.1 - 1 89
          AK096891.1 - 2 445
          AK122965.1 - 10 1787
          AK124140.1 - 17 2123
          AK223095.1 - 15 1804
          AK293852.1 - 13 1650
          AK298526.1 - 12 1413
          AK299903.1 - 14 1626
          AK307463.1 - 3 399
          AK315103.1 - 16 1969
          AL133565.1 - 14 1746
          AL833851.1 - 5 616
          BC003045.1 - 13 1628 2823536
          BC004238.2 - 13 1628 2823536
          BC028353.2 - 16 1909 4836844

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          neural tube closure
          signal transduction downstream of smoothened
          embryonic body morphogenesis
          negative regulation of smoothened signaling pathway involved in ventral spinal cord patterning
          embryonic heart tube development
          embryonic forelimb morphogenesis
          intraciliary anterograde transport
          intraciliary retrograde transport
          intraciliary transport involved in cilium morphogenesis
          cilium assembly
          embryonic digit morphogenesis
          negative regulation of smoothened signaling pathway
          camera-type eye morphogenesis
          negative regulation of epithelial cell proliferation
          limb development
          cilium morphogenesis
          ciliary receptor clustering involved in smoothened signaling pathway
          smoothened signaling pathway involved in dorsal/ventral neural tube patterning
          embryonic heart tube left/right pattern formation
          protein localization to cilium
          establishment of protein localization to organelle
          protein binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-7b8d7ffddb-2cdg5:80/100.66.130.62:80.
          git-commit: 44eaffc35a31f29f64b20d0c2dd51bfa522c97f5
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0