Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other EYA1 SIRNA Assays ›
Gene Symbol:
EYA1
Gene Name:
EYA transcriptional coactivator and phosphatase 1
Gene Aliases:
BOP, BOR, BOS1, OFC1
Chromosome Location:
Chr.8: 71197433 - 71548130 on Build GRCh38
UniGene ID:
Hs.444971
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s534824
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: EYA1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_000503.5
NP_000494.2
9
1348
NM_001288574.1
NP_001275503.1
9
1330
NM_001288575.1
NP_001275504.1
9
1266
NM_172058.3
NP_742055.1
8
940
NM_172059.3
NP_742056.1
8
747
NM_172060.3
NP_742057.1
7
812
XM_011517483.2
XP_011515785.1
8
5645
XM_011517484.2
XP_011515786.2
10
1091
XM_017013201.1
XP_016868690.1
10
1134
XM_017013202.1
XP_016868691.1
10
978
XM_017013203.1
XP_016868692.1
10
1103
XM_017013204.1
XP_016868693.1
10
1089
XM_017013205.1
XP_016868694.1
10
1106
XM_017013206.1
XP_016868695.1
11
1087
XM_017013207.1
XP_016868696.1
10
1105
XM_017013208.1
XP_016868697.1
9
1370
XM_017013209.1
XP_016868698.1
10
980
XM_017013210.1
XP_016868699.1
10
1089
XM_017013211.1
XP_016868700.1
9
978
XM_017013212.1
XP_016868701.1
8
1245
XM_017013213.1
XP_016868702.1
8
5783
GenBank mRNA
AF467247.1
-
8
722
AJ000097.1
-
8
885
AJ000098.1
-
9
1346
AK127798.1
-
9
766
AK295897.1
-
9
839
BC121798.1
-
7
1171
40084971
BC121799.1
-
8
852
40084967
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
establishment of mitotic spindle orientation
metanephros development
branching involved in ureteric bud morphogenesis
outflow tract morphogenesis
double-strand break repair
transcription, DNA-templated
pattern specification process
mesodermal cell fate specification
sensory perception of sound
anatomical structure morphogenesis
response to ionizing radiation
striated muscle tissue development
histone dephosphorylation
protein sumoylation
cellular protein localization
establishment or maintenance of apical/basal cell polarity
peptidyl-tyrosine dephosphorylation
aorta morphogenesis
outer ear morphogenesis
middle ear morphogenesis
regulation of neuron differentiation
positive regulation of DNA repair
positive regulation of Notch signaling pathway
positive regulation of transcription from RNA polymerase II promoter
neuron fate specification
embryonic skeletal system morphogenesis
semicircular canal morphogenesis
positive regulation of epithelial cell proliferation
pharyngeal system development
lung epithelial cell differentiation
otic vesicle morphogenesis
positive regulation of secondary heart field cardioblast proliferation
cochlea morphogenesis
negative regulation of extrinsic apoptotic signaling pathway in absence of ligand
RNA binding
protein tyrosine phosphatase activity
protein binding
metal ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.31.0-2025.06.23-1.0