Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other KIAA1109 SIRNA Assays ›
Gene Symbol:
KIAA1109
Gene Name:
KIAA1109
Gene Aliases:
FSA, Tweek
Chromosome Location:
Chr.4: 122152333 - 122362759 on Build GRCh38
UniGene ID:
Hs.408142
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s534978
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: KIAA1109
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_015312.3
NP_056127.2
45
8119
XM_005263282.1
XP_005263339.1
47
8447
XM_005263287.1
XP_005263344.1
47
8447
XM_006714344.1
XP_006714407.1
47
8444
XM_011532319.1
XP_011530621.1
47
8447
XM_011532320.2
XP_011530622.1
48
8542
XM_011532321.2
XP_011530623.1
46
8249
XM_011532322.1
XP_011530624.1
47
8444
XM_011532323.1
XP_011530625.1
47
8447
XM_011532324.1
XP_011530626.1
47
8447
XM_011532325.1
XP_011530627.1
47
8447
XM_011532326.1
XP_011530628.1
46
8318
XM_011532327.1
XP_011530629.1
48
8493
XM_011532330.1
XP_011530632.1
31
6393
XM_017008695.1
XP_016864184.1
47
8447
XM_017008696.1
XP_016864185.1
47
8447
XM_017008697.1
XP_016864186.1
47
8447
XM_017008698.1
XP_016864187.1
25
6409
XM_017008699.1
XP_016864188.1
47
8447
GenBank mRNA
AB029032.2
-
11
2318
AK025057.1
-
5
1326
AK095216.1
-
14
2828
AK225886.1
-
5
1274
AK309063.1
-
17
3240
AL137384.1
-
9
1946
CR936613.1
-
19
3792
DQ335469.1
-
45
8119
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
regulation of cell growth
regulation of epithelial cell differentiation
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7b7b59cd5b-lqskn:80/100.66.131.83:80.
git-commit: 87deb889a5bb48f859ebd9970e87be562fd9396c
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.32.0-2025.07.28-1.0