Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other DMPK SIRNA Assays ›
          Gene Symbol:
          DMPK
          Gene Name:
          dystrophia myotonica protein kinase
          Gene Aliases:
          DM, DM1, DM1PK, DMK, MDPK, MT-PK
          Chromosome Location:
          Chr.19: 45769709 - 45782557 on Build GRCh38
          UniGene ID:
          Hs.631596
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s536372

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: DMPK
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001081560.2 NP_001075029.1 2, 3 464
          NM_001081562.2 NP_001075031.1 2, 3 464
          NM_001081563.2 NP_001075032.1 1, 2 833
          NM_001288764.1 NP_001275693.1 3, 4 523
          NM_001288766.1 NP_001275695.1 2, 3 464
          NM_004409.4 NP_004400.4 2, 3 464
          GenBank mRNA AB209363.1 - 2, 3 1307
          AK302874.1 - 2, 3 397
          AK304749.1 - 1, 2 736
          BC026328.1 - 3, 4 518 4792104
          BC062553.1 - 2, 3 385 6480902
          BG706056.1 - 3, 4 523
          KJ534826.1 - 1, 2 289
          KJ534827.1 - 1, 2 289
          KJ534828.1 - 1, 2 289
          L19268.1 - 2, 3 386
          M87312.1 - 1, 2 834
          M94203.1 - 2, 3 136
          S72883.1 - 2, 3 295

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of sodium ion transport
          protein phosphorylation
          cellular calcium ion homeostasis
          nuclear envelope organization
          regulation of heart contraction
          muscle cell apoptotic process
          regulation of myotube differentiation
          regulation of skeletal muscle contraction by calcium ion signaling
          regulation of excitatory postsynaptic membrane potential involved in skeletal muscle contraction
          peptidyl-serine phosphorylation
          regulation of catalytic activity
          regulation of synapse structural plasticity
          regulation of cardiac conduction
          protein serine/threonine kinase activity
          protein binding
          ATP binding
          myosin phosphatase regulator activity
          heat shock protein binding
          metal ion binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-865d7754c7-mcq8j:80/100.66.130.62:80.
          git-commit: 7ff36f61b459eb8f02ba5a263f0e3ce367ca82d3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0