Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other TCTN1 SIRNA Assays ›
          Gene Symbol:
          TCTN1
          Gene Name:
          tectonic family member 1
          Gene Aliases:
          JBTS13, TECT1
          Chromosome Location:
          Chr.12: 110614027 - 110649130 on Build GRCh38
          UniGene ID:
          Hs.211511
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s536490

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: TCTN1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001082537.2 NP_001076006.1 3, 4 639
          NM_001082538.2 NP_001076007.1 3, 4 639
          NM_001173975.2 NP_001167446.1 3, 4 700
          NM_001173976.1 NP_001167447.1 4, 5 715
          NM_001319680.1 NP_001306609.1 3, 4 639
          NM_001319681.1 NP_001306610.1 4, 5 812
          NM_001319682.1 NP_001306611.1 3, 4 700
          NM_024549.5 NP_078825.2 3, 4 639
          NR_135088.1 4, 5 995
          XM_005253934.3 XP_005253991.1 3, 4 543
          XM_005253935.3 XP_005253992.1 3, 4 543
          XM_006719594.2 XP_006719657.1 3, 4 604
          XM_006719595.2 XP_006719658.1 4, 5 710
          XM_006719596.2 XP_006719659.1 4, 5 733
          XM_006719597.3 XP_006719660.1 2, 3 430
          XM_011538733.2 XP_011537035.1 3, 4 543
          XM_011538734.2 XP_011537036.1 2, 3 498
          XM_011538735.1 XP_011537037.1 3, 4 543
          XM_011538737.2 XP_011537039.1 3, 4 543
          XM_011538738.2 XP_011537040.1 3, 4 543
          XM_017019964.1 XP_016875453.1 3, 4 604
          XM_017019965.1 XP_016875454.1 4, 5 822
          XM_017019966.1 XP_016875455.1 4, 5 869
          XM_017019967.1 XP_016875456.1 4, 5 577
          XM_017019968.1 XP_016875457.1 3, 4 666
          GenBank mRNA AK024780.1 - 4, 5 613
          AK055891.1 - 3, 4 553
          AK092775.1 - 3, 4 601
          AK295514.1 - 3, 4 639
          AK301732.1 - 4, 5 677
          AY358184.1 - 3, 4 502
          BC033811.1 - 3, 4 586
          BC040113.1 - 3, 4 529 6054789
          BC044885.1 - 4, 5 857
          BC062611.1 - 3, 4 490 5184202
          DN997186.1 - 3, 4 599
          DQ278868.1 - 3, 4 564

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s)

          in utero embryonic development
          neural tube formation
          regulation of smoothened signaling pathway
          somatic motor neuron differentiation
          telencephalon development
          dorsal/ventral neural tube patterning
          central nervous system interneuron axonogenesis
          cilium morphogenesis

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0