Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other TCTN1 SIRNA Assays ›
          Gene Symbol:
          TCTN1
          Gene Name:
          tectonic family member 1
          Gene Aliases:
          JBTS13, TECT1
          Chromosome Location:
          Chr.12: 110614027 - 110649130 on Build GRCh38
          UniGene ID:
          Hs.211511
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s536491

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: TCTN1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001082537.2 NP_001076006.1 6 901
          NM_001082538.2 NP_001076007.1 6 901
          NM_001173975.2 NP_001167446.1 6 962
          NM_001173976.1 NP_001167447.1 7 977
          NM_001319680.1 NP_001306609.1 6 901
          NM_001319681.1 NP_001306610.1 7 1074
          NR_135088.1 7 1257
          XM_005253934.3 XP_005253991.1 6 805
          XM_005253935.3 XP_005253992.1 6 805
          XM_006719594.2 XP_006719657.1 6 866
          XM_006719595.2 XP_006719658.1 7 972
          XM_006719596.2 XP_006719659.1 7 995
          XM_006719597.3 XP_006719660.1 5 692
          XM_006719598.2 XP_006719661.1 7 965
          XM_006719599.2 XP_006719662.1 6 770
          XM_006719600.2 XP_006719663.1 5 709
          XM_011538734.2 XP_011537036.1 5 760
          XM_011538735.1 XP_011537037.1 6 805
          XM_011538737.2 XP_011537039.1 6 805
          XM_011538738.2 XP_011537040.1 6 805
          XM_017019964.1 XP_016875453.1 6 866
          XM_017019965.1 XP_016875454.1 7 1084
          XM_017019966.1 XP_016875455.1 7 1131
          XM_017019967.1 XP_016875456.1 7 839
          XM_017019968.1 XP_016875457.1 6 928
          GenBank mRNA AK024780.1 - 7 875
          AK092775.1 - 6 863
          AK301732.1 - 7 939
          AY358184.1 - 6 764
          BC044885.1 - 7 1119
          BC062611.1 - 6 752 5184202
          DQ278868.1 - 6 826

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s)

          in utero embryonic development
          neural tube formation
          regulation of smoothened signaling pathway
          somatic motor neuron differentiation
          telencephalon development
          dorsal/ventral neural tube patterning
          central nervous system interneuron axonogenesis
          cilium morphogenesis

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0