Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CEP290 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025114.3 | NP_079390.3 | 25 | 3070 | ||
XM_011538756.2 | XP_011537058.1 | 27 | 3931 | |||
XM_011538757.2 | XP_011537059.1 | 27 | 3739 | |||
XM_011538758.2 | XP_011537060.1 | 27 | 3931 | |||
XM_011538759.2 | XP_011537061.1 | 27 | 3931 | |||
XM_011538760.1 | XP_011537062.1 | 27 | 3931 | |||
XM_011538761.1 | XP_011537063.1 | 27 | 3931 | |||
XM_011538762.2 | XP_011537064.1 | 26 | 3163 | |||
XM_011538763.2 | XP_011537065.1 | 25 | 3070 | |||
XM_011538764.2 | XP_011537066.1 | 27 | 3931 | |||
XM_011538765.2 | XP_011537067.1 | 27 | 3931 | |||
XM_011538766.2 | XP_011537068.1 | 13 | 2158 | |||
XM_017019980.1 | XP_016875469.1 | 27 | 3931 | |||
XM_017019981.1 | XP_016875470.1 | 27 | 3931 | |||
XM_017019982.1 | XP_016875471.1 | 27 | 3931 | |||
XM_017019983.1 | XP_016875472.1 | 24 | 3049 | |||
GenBank mRNA | AK025632.1 | - | 13 | 1673 | ||
AY590151.1 | - | 17 | 1991 | |||
BC008641.1 | - | 1 | 122 | |||
BC043398.1 | - | 25 | 2825 | |||
DQ109808.1 | - | 25 | 3070 |