Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FUT3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000149.3 | NP_000140.1 | 3 | 1041 | ||
NM_001097639.1 | NP_001091108.1 | 3 | 693 | |||
NM_001097640.1 | NP_001091109.1 | 3 | 639 | |||
NM_001097641.1 | NP_001091110.1 | 2 | 500 | |||
XM_011527865.1 | XP_011526167.1 | 3 | 1019 | |||
XM_011527866.1 | XP_011526168.1 | 3 | 680 | |||
XM_011527867.1 | XP_011526169.1 | 3 | 649 | |||
GenBank mRNA | AF131913.1 | - | 2 | 572 | ||
AK291852.1 | - | 3 | 668 | |||
AK313575.1 | - | 3 | 690 | |||
BC074836.2 | - | 2 | 445 | 30915328 | ||
BC074837.2 | - | 2 | 445 | 30915548 | ||
BC108675.1 | - | 2 | 497 | 5187745 | ||
U27326.1 | - | 3 | 632 | |||
U27327.1 | - | 3 | 686 | |||
U27328.1 | - | 4 | 1035 | |||
X53578.1 | - | 2 | 475 |