Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MINOS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001032363.3 | NP_001027535.1 | 3 | 257 | ||
NM_001204082.1 | NP_001191011.1 | 2 | 209 | |||
NM_001204083.1 | NP_001191012.1 | 3 | 257 | |||
NR_033757.2 | 4 | 442 | ||||
NR_033758.2 | 4 | 336 | ||||
GenBank mRNA | AJ711911.1 | - | 4 | 333 | ||
AK094318.1 | - | 3 | 234 | |||
BC009927.1 | - | 4 | 312 | |||
BC070388.1 | - | 3 | 244 | 30407082 | ||
BG687187.1 | - | 265 | ||||
BM784596.1 | - | 2 | 162 | |||
CB157536.1 | - | 3 | 193 | |||
DA523181.1 | - | 3 | 258 | |||
DA793500.1 | - | 4 | 399 |