Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM21A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005751.2 | NP_001005751.1 | 30 | 3940 | ||
NM_001291398.1 | NP_001278327.1 | 29 | 3877 | |||
XM_005269806.3 | XP_005269863.2 | 29 | 3816 | |||
XM_005269807.3 | XP_005269864.2 | 29 | 3801 | |||
XM_005269808.3 | XP_005269865.2 | 28 | 3702 | |||
XM_005269810.3 | XP_005269867.2 | 12 | 2033 | |||
XM_006717831.3 | XP_006717894.2 | 29 | 3735 | |||
XM_006717832.3 | XP_006717895.2 | 27 | 3600 | |||
XM_011539786.2 | XP_011538088.1 | 27 | 3489 | |||
XM_017016227.1 | XP_016871716.1 | 29 | 3765 | |||
XM_017016228.1 | XP_016871717.1 | 28 | 3753 | |||
XM_017016229.1 | XP_016871718.1 | 28 | 3738 | |||
XM_017016230.1 | XP_016871719.1 | 29 | 3753 | |||
XM_017016231.1 | XP_016871720.1 | 28 | 3672 | |||
XM_017016232.1 | XP_016871721.1 | 27 | 3666 | |||
XM_017016233.1 | XP_016871722.1 | 27 | 3463 | |||
XM_017016234.1 | XP_016871723.1 | 29 | 3823 | |||
XM_017016235.1 | XP_016871724.1 | 21 | 2839 | |||
GenBank mRNA | AK125147.1 | - | 2 | 2294 | ||
AK304116.1 | - | 30 | 3876 | |||
AL080183.1 | - | 6 | 1272 | |||
BC065500.1 | - | 2 | 178 | |||
BC075815.1 | - | 26 | 3442 | |||
BC082258.1 | - | 30 | 3867 | 6278433 | ||
BC144479.1 | - | 29 | 3906 | 9053006 | ||
BQ060891.1 | - | 2 | 700 |