Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GXYLT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080393.1 | NP_001073862.1 | 4 | 827 | ||
XM_011534067.2 | XP_011532369.1 | 4 | 693 | |||
XM_011534068.1 | XP_011532370.1 | 4 | 712 | |||
XM_017007124.1 | XP_016862613.1 | 4 | 697 | |||
GenBank mRNA | BC127733.1 | - | 3 | 473 | 40127813 |