Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other APP SIRNA Assays ›
          Gene Symbol:
          App
          Gene Name:
          amyloid beta (A4) precursor protein
          Gene Aliases:
          Abeta, Abpp, Adap, Ag, Cvap, E030013M08Rik, betaApp
          Chromosome Location:
          Chr.16: 84954436 - 85173952 on Build GRCm38
          UniGene ID:
          Mm.277585
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID s62514

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: App
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001198823.1 NP_001185752.1 16 2147
          NM_001198824.1 NP_001185753.1 15 2090
          NM_001198825.1 NP_001185754.1 14 2036
          NM_001198826.1 NP_001185755.1 15 2093
          NM_007471.3 NP_031497.2 14 1922
          GenBank mRNA AK052448.1 - 4 343
          AK076506.1 - 8 842
          AK128971.1 - 14 2045
          AK133639.1 - 15 2087
          AK147543.1 - 14 1958
          AK148439.1 - 15 1966
          AK159245.1 - 15 2102
          AK159718.1 - 15 2079
          AK161854.1 - 9 1003
          AK161976.1 - 14 1897
          AK190051.1 - 3 288
          AK191213.1 - 4 352
          AK191703.1 - 4 341
          AK191711.1 - 3 291
          AK192103.1 - 3 287
          AK192320.1 - 3 286
          AK192899.1 - 3 287
          AK192938.1 - 3 287
          AK194133.1 - 2 288
          AK197763.1 - 4 345
          AK198568.1 - 3 287
          AK199923.1 - 3 287
          AK200960.1 - 4 342
          AK202486.1 - 3 289
          AK202618.1 - 3 287
          AK216049.1 - 3 287
          AK216591.1 - 3 287
          AY267348.1 - 16 1998
          BC005490.1 - 12 1511
          BC070409.1 - 14 1918 5708093
          M18373.1 - 14 1911
          U84012.1 - 14 1911
          X59379.1 - 14 1841
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
          GenBankmRNA AK084102.1 - 1337 Chr. - - [-] on Build

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          suckling behavior
          mRNA polyadenylation
          regulation of translation
          protein phosphorylation
          cellular copper ion homeostasis
          endocytosis
          apoptotic process
          response to oxidative stress
          cell adhesion
          regulation of epidermal growth factor-activated receptor activity
          Notch signaling pathway
          nervous system development
          axonogenesis
          mating behavior
          locomotory behavior
          axo-dendritic transport
          cholesterol metabolic process
          adult locomotory behavior
          visual learning
          negative regulation of peptidase activity
          regulation of gene expression
          negative regulation of endopeptidase activity
          positive regulation of peptidase activity
          positive regulation of G2/M transition of mitotic cell cycle
          axon midline choice point recognition
          neuron remodeling
          dendrite development
          extracellular matrix organization
          forebrain development
          neuron projection development
          ionotropic glutamate receptor signaling pathway
          regulation of multicellular organism growth
          regulation of protein binding
          negative regulation of neuron differentiation
          positive regulation of mitotic cell cycle
          positive regulation of transcription from RNA polymerase II promoter
          collateral sprouting in absence of injury
          regulation of synapse structure or activity
          neuromuscular process controlling balance
          synaptic growth at neuromuscular junction
          protein homooligomerization
          neuron apoptotic process
          smooth endoplasmic reticulum calcium ion homeostasis
          DNA binding
          serine-type endopeptidase inhibitor activity
          receptor binding
          protein binding
          heparin binding
          peptidase activator activity
          enzyme binding
          peptidase inhibitor activity
          acetylcholine receptor binding
          identical protein binding
          metal ion binding
          transition metal ion binding
          PTB domain binding
          growth factor receptor binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0