Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: App | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001198823.1 | NP_001185752.1 | 18 | 2419 | ||
NM_001198824.1 | NP_001185753.1 | 17 | 2362 | |||
NM_001198825.1 | NP_001185754.1 | 16 | 2308 | |||
NM_001198826.1 | NP_001185755.1 | 17 | 2365 | |||
NM_007471.3 | NP_031497.2 | 16 | 2194 | |||
GenBank mRNA | AK052448.1 | - | 6 | 615 | ||
AK076506.1 | - | 10 | 1114 | |||
AK128971.1 | - | 16 | 2317 | |||
AK133639.1 | - | 17 | 2359 | |||
AK147543.1 | - | 16 | 2230 | |||
AK159245.1 | - | 17 | 2374 | |||
AK159718.1 | - | 17 | 2351 | |||
AK161854.1 | - | 11 | 1275 | |||
AK161976.1 | - | 16 | 2169 | |||
AY267348.1 | - | 18 | 2270 | |||
BC005490.1 | - | 14 | 1783 | |||
BC070409.1 | - | 16 | 2190 | 5708093 | ||
M18373.1 | - | 16 | 2183 | |||
U84012.1 | - | 16 | 2183 | |||
X59379.1 | - | 16 | 2113 |