Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arhgap6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001287530.1 | NP_001274459.1 | 8 | 1276 | ||
NM_009707.4 | NP_033837.2 | 8 | 2644 | |||
NM_178754.3 | NP_848869.1 | 8 | 1187 | |||
GenBank mRNA | AF012273.2 | - | 1766 | |||
AF177664.1 | - | 1766 | ||||
AK048162.1 | - | 8 | 1141 | |||
AK048507.1 | - | 8 | 1142 | |||
AK077996.1 | - | 8 | 1149 | |||
AK079651.1 | - | 8 | 1100 | |||
AK166015.1 | - | 8 | 1107 | |||
AK170575.1 | - | 8 | 1143 |