Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Btg3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001297747.1 | NP_001284676.1 | 4 | 731 | ||
NM_009770.3 | NP_033900.1 | 5 | 927 | |||
GenBank mRNA | AK144415.1 | - | 5 | 878 | ||
BC012705.1 | - | 5 | 866 | 3980599 | ||
BC094027.1 | - | 5 | 882 | 4457150 | ||
BC147656.1 | - | 4 | 731 | 9087445 | ||
BC147662.1 | - | 4 | 731 | 9087451 | ||
D83745.1 | - | 5 | 884 | |||
Z72000.1 | - | 5 | 902 |
Gene Symbol: Gm7334 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AA265715.1 | - | 1 | 117 |