Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Camk2g | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039138.2 | NP_001034227.1 | 2 | 246 | ||
NM_001039139.2 | NP_001034228.1 | 2 | 246 | |||
NM_178597.5 | NP_848712.2 | 2 | 246 | |||
GenBank mRNA | AF395884.1 | - | 2 | 123 | ||
AK031213.1 | - | 2 | 209 | |||
AK054406.1 | - | 2 | 213 | |||
AK078311.1 | - | 2 | 190 | |||
AK081969.1 | - | 2 | 208 | |||
AK154764.1 | - | 2 | 175 | |||
BC019162.1 | - | 2 | 198 | 3675224 | ||
BC025597.1 | - | 2 | 200 | 5342641 | ||
CJ151263.1 | - | 2 | 246 |