Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cd34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001111059.1 | NP_001104529.1 | 2, 3 | 400 | ||
NM_133654.3 | NP_598415.1 | 2, 3 | 400 | |||
GenBank mRNA | AK044121.1 | - | 2, 3 | 336 | ||
AK051507.1 | - | 2 | 1614 | |||
AK053874.1 | - | 2, 3 | 336 | |||
AK089010.1 | - | 2, 3 | 306 | |||
AK167329.1 | - | 2, 3 | 311 | |||
AK167350.1 | - | 2, 3 | 311 | |||
BC006607.1 | - | 2, 3 | 316 | 3966337 | ||
BC066820.1 | - | 2, 3 | 273 | |||
BY249301.1 | - | 2, 3 | 403 | |||
S69293.1 | - | 2, 3 | 278 | |||
S69299.1 | - | 2, 3 | 255 | |||
S69301.1 | - | 2, 3 | 255 |