Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdk4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009870.3 | NP_034000.1 | 3 | 436 | ||
GenBank mRNA | AK003804.1 | - | 3 | 403 | ||
AK011871.1 | - | 3 | 424 | |||
AK019380.1 | - | 3 | 390 | |||
AK078440.1 | - | 3 | 390 | |||
AK146107.1 | - | 3 | 403 | |||
AK153465.1 | - | 3 | 436 | |||
AK185090.1 | - | 3 | 328 | |||
BC031599.1 | - | 3 | 425 | |||
BC046336.1 | - | 3 | 436 | 6312184 | ||
BC052694.1 | - | 3 | 431 | 5714971 | ||
L01640.1 | - | 2 | 263 |