Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cflar | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289704.2 | NP_001276633.1 | 4 | 670 | ||
NM_001293804.1 | NP_001280733.1 | 4 | 653 | |||
NM_001293805.1 | NP_001280734.1 | 2 | 146 | |||
NM_009805.4 | NP_033935.2 | 4 | 942 | |||
NM_207653.5 | NP_997536.1 | 4 | 942 | |||
GenBank mRNA | AK143812.1 | - | 4 | 935 | ||
AK152515.1 | - | 4 | 637 | |||
AK161120.1 | - | 2 | 149 | |||
BC029223.1 | - | 4 | 911 | 4460554 | ||
BC085105.1 | - | 4 | 665 | |||
U97076.1 | - | 4 | 608 | |||
Y14041.1 | - | 3 | 520 | |||
Y14042.1 | - | 3 | 520 |