Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ctsb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007798.3 | NP_031824.1 | 3 | 275 | ||
GenBank mRNA | AK002456.1 | - | 3 | 224 | ||
AK083393.1 | - | 3 | 275 | |||
AK147192.1 | - | 3 | 222 | |||
AK149884.1 | - | 3 | 224 | |||
AK151790.1 | - | 3 | 222 | |||
AK153047.1 | - | 3 | 221 | |||
AK159992.1 | - | 3 | 260 | |||
AK167361.1 | - | 3 | 256 | |||
AK170959.1 | - | 3 | 248 | |||
AK190209.1 | - | 2 | 73 | |||
AK191055.1 | - | 2 | 73 | |||
AK191391.1 | - | 2 | 73 | |||
AK196955.1 | - | 2 | 73 | |||
AK198213.1 | - | 2 | 73 | |||
AK199268.1 | - | 2 | 74 | |||
AK202291.1 | - | 2 | 73 | |||
AK203639.1 | - | 2 | 73 | |||
BC006656.1 | - | 3 | 266 | 3500700 | ||
M14222.1 | - | 2 | 180 | |||
S69034.1 | - | 3 | 271 |