Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other CTSD SIRNA Assays ›
          Gene Symbol:
          Ctsd
          Gene Name:
          cathepsin D
          Gene Aliases:
          CD, CatD
          Chromosome Location:
          Chr.7: 142375916 - 142387870 on Build GRCm38
          UniGene ID:
          Mm.231395
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID s64603

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Ctsd
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_009983.2 NP_034113.1 5, 6 757
          GenBank mRNA AK089294.1 - 5, 6 715
          AK150363.1 - 5, 6 718
          AK150364.1 - 5, 6 715
          AK150579.1 - 5, 6 715
          AK150622.1 - 5, 6 715
          AK150636.1 - 5, 6 715
          AK150694.1 - 5, 6 715
          AK150768.1 - 5, 6 715
          AK150946.1 - 5, 6 767
          AK151256.1 - 5, 6 715
          AK151275.1 - 5, 6 715
          AK151276.1 - 5, 6 715
          AK151374.1 - 5, 6 715
          AK151829.1 - 5, 6 715
          AK151962.1 - 5, 6 718
          AK152036.1 - 5, 6 728
          AK152045.1 - 5, 6 768
          AK152168.1 - 5, 6 715
          AK152187.1 - 5, 6 715
          AK152188.1 - 5, 6 715
          AK152406.1 - 5, 6 718
          AK152458.1 - 5, 6 715
          AK152567.1 - 5, 6 715
          AK152578.1 - 5, 6 715
          AK152593.1 - 5, 6 745
          AK152638.1 - 5, 6 745
          AK152680.1 - 5, 6 715
          AK152828.1 - 5, 6 715
          AK152881.1 - 5, 6 715
          AK152892.1 - 5, 6 715
          AK153191.1 - 5, 6 715
          AK153291.1 - 5, 6 715
          AK159097.1 - 5, 6 715
          AK159209.1 - 5, 6 715
          AK159440.1 - 5, 6 768
          AK159490.1 - 5, 6 715
          AK159512.1 - 5, 6 757
          AK159580.1 - 5, 6 715
          AK159642.1 - 3, 4 467
          AK159743.1 - 5, 6 719
          AK159896.1 - 5, 6 715
          AK159970.1 - 5, 6 768
          AK165227.1 - 5, 6 785
          AK166864.1 - 5, 6 728
          AK166938.1 - 5, 6 762
          AK167143.1 - 5, 6 753
          AK167366.1 - 5, 6 787
          AK167698.1 - 5, 6 714
          AK167750.1 - 5, 6 728
          AK167867.1 - 5, 6 716
          AK169139.1 - 5, 6 768
          AK171195.1 - 5, 6 716
          AK172583.1 - 5, 6 715
          BC019682.1 - 1, 2 112
          BC054758.1 - 5, 6 787 6413315
          BC057931.1 - 5, 6 748 5253319
          X52886.1 - 5, 6 757
          X53337.1 - 5, 6 697

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: NM_134334, X54467, BC062032, FQ228180, FQ232703.

          Gene Ontology Categories:

          Function(s) Process(es)

          autophagosome assembly
          proteolysis
          autophagy
          protein catabolic process
          epithelial tube branching involved in lung morphogenesis
          endopeptidase activity
          aspartic-type endopeptidase activity
          peptidase activity
          hydrolase activity
          peptide binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-6467d75d44-xp7n2:80/100.66.130.62:80.
          git-commit: 8ea1bebb48d33b8b7c0e2103582aa482e6db8664
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0