Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cttn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252572.1 | NP_001239501.1 | 5, 6 | 625 | ||
NM_007803.5 | NP_031829.2 | 5, 6 | 625 | |||
GenBank mRNA | AK084249.1 | - | 5, 6 | 435 | ||
AK146856.1 | - | 5, 6 | 432 | |||
AK148010.1 | - | 5, 6 | 428 | |||
AK165555.1 | - | 5, 6 | 428 | |||
AK168699.1 | - | 5, 6 | 426 | |||
AK168871.1 | - | 5, 6 | 426 | |||
AK168945.1 | - | 5, 6 | 436 | |||
AK197295.1 | - | 1, 2 | 30 | |||
BC011434.1 | - | 5, 6 | 418 | 3981559 | ||
BC117820.1 | - | 4, 5 | 326 | |||
U03184.1 | - | 5, 6 | 414 |