Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ddx3x | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010028.3 | NP_034158.1 | 12 | 1392 | ||
GenBank mRNA | AK140990.1 | - | 12 | 1393 | ||
AK163250.1 | - | 12 | 1367 | |||
AK176969.1 | - | 2 | 175 | |||
AK186622.1 | - | 2 | 176 | |||
AK204341.1 | - | 2 | 213 | |||
AK205639.1 | - | 2 | 176 | |||
BC150862.1 | - | 12 | 1346 | 9087773 | ||
BC172016.1 | - | 12 | 1343 | 9054302 | ||
DQ858457.1 | - | 2 | 181 | |||
DQ858458.1 | - | 2 | 181 | |||
L25126.1 | - | 12 | 1324 | |||
Z38117.1 | - | 12 | 1376 |