Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ednrb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136061.2 | NP_001129533.1 | 5 | 1340 | ||
NM_001276296.1 | NP_001263225.1 | 4 | 1154 | |||
NM_007904.4 | NP_031930.1 | 4 | 1154 | |||
GenBank mRNA | AK076426.1 | - | 4 | 1148 | ||
AK082103.1 | - | 5 | 1141 | |||
AK083415.1 | - | 5 | 1037 | |||
AK085165.1 | - | 4 | 1146 | |||
AK085532.1 | - | 4 | 1150 | |||
AK190192.1 | - | 1 | 15 | |||
AK198545.1 | - | 1 | 15 | |||
AK211542.1 | - | 1 | 15 | |||
BC026553.1 | - | 5 | 1266 | 4971909 | ||
U32329.1 | - | 4 | 1132 |