Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eif4a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159375.1 | NP_001152847.1 | 2, 3 | 200 | ||
NM_144958.4 | NP_659207.1 | 2, 3 | 200 | |||
GenBank mRNA | AK010644.1 | - | 2, 3 | 102 | ||
AK011529.1 | - | 2, 3 | 93 | |||
AK077429.1 | - | 2, 3 | 93 | |||
AK088124.1 | - | 2, 3 | 95 | |||
AK135752.1 | - | 2, 3 | 93 | |||
AK135994.1 | - | 2, 3 | 94 | |||
AK144850.1 | - | 2, 3 | 93 | |||
AK145375.1 | - | 2, 3 | 95 | |||
AK145508.1 | - | 2, 3 | 95 | |||
AK146424.1 | - | 2, 3 | 95 | |||
AK146464.1 | - | 2, 3 | 93 | |||
AK152694.1 | - | 2, 3 | 95 | |||
AK159643.1 | - | 2, 3 | 95 | |||
AK162017.1 | - | 2, 3 | 95 | |||
AK166439.1 | - | 2, 3 | 93 | |||
AK166527.1 | - | 2, 3 | 93 | |||
AK166572.1 | - | 2, 3 | 95 | |||
AK166674.1 | - | 2, 3 | 95 | |||
AK167003.1 | - | 2, 3 | 93 | |||
AK167083.1 | - | 2, 3 | 93 | |||
AK167196.1 | - | 2, 3 | 95 | |||
AK167205.1 | - | 2, 3 | 92 | |||
AK167906.1 | - | 2, 3 | 95 | |||
AK167988.1 | - | 2, 3 | 93 | |||
AK168697.1 | - | 2, 3 | 94 | |||
AK168780.1 | - | 2, 3 | 93 | |||
AK169023.1 | - | 2, 3 | 93 | |||
AK169030.1 | - | 2, 3 | 86 | |||
AK169057.1 | - | 2, 3 | 93 | |||
AK169156.1 | - | 2, 3 | 93 | |||
AK169279.1 | - | 2, 3 | 93 | |||
AK179026.1 | - | 1, 2 | 23 | |||
AK182003.1 | - | 23 | ||||
AK207714.1 | - | 1, 2 | 23 | |||
AK210015.1 | - | 1, 2 | 23 | |||
AK211426.1 | - | 23 | ||||
BC049915.1 | - | 2, 3 | 83 | 5068709 | ||
BC099392.1 | - | 1, 2 | 76 | |||
X03039.1 | - | 2, 3 | 90 | |||
X03040.1 | - | 1, 2 | 93 |