Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eln | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007925.3 | NP_031951.2 | 26 | 1799 | ||
GenBank mRNA | AK041860.1 | - | 26 | 1800 | ||
AK080277.1 | - | 26 | 1828 | |||
BC034057.1 | - | 14 | 1337 | |||
BC051074.1 | - | 14 | 873 | |||
BC051649.1 | - | 26 | 1868 | 5346255 | ||
U08210.1 | - | 26 | 1786 |