Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gart | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010256.2 | NP_034386.2 | 14 | 1771 | ||
GenBank mRNA | AK146355.1 | - | 14 | 1756 | ||
AK146478.1 | - | 14 | 1713 | |||
AK168458.1 | - | 14 | 1731 | |||
AK168501.1 | - | 14 | 1734 | |||
AK168724.1 | - | 14 | 1746 | |||
AK168796.1 | - | 14 | 1746 | |||
AK168864.1 | - | 14 | 1722 | |||
AK168876.1 | - | 14 | 1723 | |||
AK181084.1 | - | 3 | 319 | |||
AK184743.1 | - | 3 | 318 | |||
AK212300.1 | - | 3 | 318 | |||
BC070465.1 | - | 14 | 1718 | 30611835 | ||
U01024.1 | - | 13 | 1676 |