Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hipk3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145824.1 | NP_001139296.1 | 16 | 3898 | ||
NM_010434.2 | NP_034564.2 | 16 | 3901 | |||
GenBank mRNA | AF077660.1 | - | 16 | 3655 | ||
AF170305.1 | - | 15 | 3365 | |||
AF305238.1 | - | 15 | 3363 | |||
AK140130.1 | - | 16 | 4004 | |||
BC131663.1 | - | 16 | 3533 | |||
BC131664.1 | - | 16 | 3529 | 40110751 |