Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ifi203 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF022371.1 | - | 373 | |||
AK040214.1 | - | 3 | 313 | |||
AK041232.1 | - | 3 | 301 | |||
AK137616.1 | - | 3 | 312 | |||
AK144663.1 | - | 3 | 294 | |||
AK169754.1 | - | 3 | 307 | |||
AK172303.1 | - | 366 | ||||
AK172505.1 | - | 387 | ||||
BC008167.1 | - | 415 | 3257492 | |||
BE632791.1 | - | 1 | 91 | |||
CA574467.1 | - | 1 | 29 |
Gene Symbol: Mndal | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170853.1 | NP_001164324.1 | 312 | |||
GenBank mRNA | AK087931.1 | - | 1 | 141 | ||
AK136391.1 | - | 313 | ||||
FJ594740.1 | - | 1 | 211 | |||
FJ594741.1 | - | 220 |