Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lox | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286181.1 | NP_001273110.1 | 4 | 897 | ||
NM_001286182.1 | NP_001273111.1 | 4 | 854 | |||
NM_010728.3 | NP_034858.2 | 3 | 1274 | |||
GenBank mRNA | AK135213.1 | - | 3 | 405 | ||
AK141995.1 | - | 3 | 1160 | |||
AK159199.1 | - | 3 | 1077 | |||
AK159241.1 | - | 3 | 1053 | |||
AK159263.1 | - | 3 | 1114 | |||
AK159368.1 | - | 3 | 1036 | |||
AK159682.1 | - | 3 | 987 | |||
AK159796.1 | - | 3 | 1048 | |||
AK160096.1 | - | 3 | 1048 | |||
AK164634.1 | - | 4 | 855 | |||
AK166279.1 | - | 3 | 982 | |||
AK166386.1 | - | 1118 | ||||
AK166442.1 | - | 1274 | ||||
BC018439.1 | - | 3 | 1125 | 2655752 | ||
M65142.1 | - | 3 | 1042 |