Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lox | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286181.1 | NP_001273110.1 | 6 | 1154 | ||
NM_001286182.1 | NP_001273111.1 | 6 | 1111 | |||
NM_010728.3 | NP_034858.2 | 5 | 1531 | |||
GenBank mRNA | AK135213.1 | - | 5 | 662 | ||
AK141995.1 | - | 5 | 1417 | |||
AK159199.1 | - | 5 | 1334 | |||
AK159241.1 | - | 5 | 1310 | |||
AK159263.1 | - | 5 | 1371 | |||
AK159368.1 | - | 5 | 1293 | |||
AK159682.1 | - | 5 | 1244 | |||
AK159796.1 | - | 5 | 1305 | |||
AK160096.1 | - | 5 | 1305 | |||
AK166279.1 | - | 5 | 1239 | |||
AK166386.1 | - | 1375 | ||||
AK166442.1 | - | 1531 | ||||
BC018439.1 | - | 5 | 1382 | 2655752 | ||
M65142.1 | - | 5 | 1299 | |||
M65143.1 | - | 2 | 158 |