Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ndst2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010811.2 | NP_034941.2 | 7 | 2231 | ||
GenBank mRNA | AF074925.1 | - | 5 | 1481 | ||
AK150100.1 | - | 7 | 2012 | |||
AK170298.1 | - | 7 | 2014 | |||
AK182571.1 | - | 2 | 77 | |||
AK199361.1 | - | 3 | 284 | |||
AK199550.1 | - | 1 | 37 | |||
BC050829.1 | - | 7 | 1996 | |||
BC110479.1 | - | 5 | 1554 | 40048114 | ||
BC110480.1 | - | 5 | 1629 | 40048115 | ||
U02304.1 | - | 6 | 1993 | |||
X75885.1 | - | 5 | 1635 |