Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sept2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159717.1 | NP_001153189.1 | 9, 10 | 932 | ||
NM_001159718.1 | NP_001153190.1 | 8, 9 | 905 | |||
NM_001159719.1 | NP_001153191.1 | 9, 10 | 980 | |||
NM_010891.2 | NP_035021.1 | 9, 10 | 936 | |||
GenBank mRNA | AK028072.1 | - | 9, 10 | 918 | ||
AK146616.1 | - | 9, 10 | 933 | |||
AK151591.1 | - | 9, 10 | 920 | |||
AK169852.1 | - | 8, 9 | 954 | |||
AK171331.1 | - | 9, 10 | 915 | |||
AK196297.1 | - | 2, 3 | 259 | |||
AK216929.1 | - | 2, 3 | 259 | |||
BC138636.1 | - | 8, 9 | 873 | 8861656 | ||
BC138637.1 | - | 8, 9 | 873 | 8861657 | ||
D49382.2 | - | 9, 10 | 891 |