Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nrxn3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001198587.3 | NP_001185516.2 | 15 | 4071 | ||
NM_001252074.2 | NP_001239003.1 | 2 | 1286 | |||
NM_172544.3 | NP_766132.2 | 15 | 2988 | |||
GenBank mRNA | AK032126.1 | - | 15 | 2980 | ||
AK042730.1 | - | 2 | 1288 | |||
AK129205.1 | - | 14 | 2861 | |||
BC060719.1 | - | 16 | 3646 | 6406001 |